Assay Details
Target Gene Details
Entrez Gene ID: | 622675 |
Gene Name: | zinc finger protein 827 |
Gene Aliases: |
2810449M09Rik, D630040G17Rik, Znf827 |
Location: |
Chr.8:79027983-79193768 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp827 | NM_001294279.1 | 15 | 6923 | NP_001281208.1 |
| NM_178267.3 | 15 | 6918 | NP_839998.2 | |
| AK033589.1 | 1 | 2775 | ||
| AK035374.1 | 1 | 1661 | ||
| AK052736.1 | 10 | 3029 | BAC35124.1 | |
| BC088739.1 | 10 | 4554 | AAH88739.1 |
More Information
Set Membership: |
|

Genomic Map