Assay Details
Target Gene Details
Entrez Gene ID: | 101994 |
Gene Name: | chromosome alignment maintaining phosphoprotein 1 |
Gene Aliases: |
AA675043, AI116001, D8Ertd457e, D8Ertd569e, Zfp828, Znf828, mKIAA1802 |
Location: |
Chr.8:13869641-13881639 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Champ1 | NM_181854.2 | 2 | 2088 | NP_862902.1 |
| AK129444.1 | 2 | 1988 | BAC98254.1 | |
| AK132164.1 | 2 | 2051 | BAE21006.1 | |
| AK133990.1 | 2 | 2014 | BAE21972.1 | |
| AK139963.1 | 2 | 2053 | ||
| AK170835.1 | 2 | 2031 | BAE42063.1 | |
| BC028991.1 | 2 | 2000 | AAH28991.1 |

Genomic Map