Assay Details
Target Gene Details
Entrez Gene ID: | 244551 |
Gene Name: | nanos homolog 3 (Drosophila) |
Gene Aliases: |
nos3 |
Location: |
Chr.8:84173730-84176945 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nanos3 | NM_194059.2 | 1 | 253 | NP_918948.1 |
| AB095973.1 | 1 | 253 | BAC82558.1 | |
| BC119513.1 | 1 | 253 | ||
| BC119514.1 | 1 | 253 |
More Information
Set Membership: |
|

Genomic Map