Assay Details
Target Gene Details
Entrez Gene ID: | 12797 |
Gene Name: | calponin 1 |
Gene Aliases: |
CN, CnnI |
Location: |
Chr.9:22099221-22109221 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnn1 | NM_009922.4 | 7 | 1158 | NP_034052.3 |
| AK035548.1 | 7 | 1131 | BAC29100.1 | |
| BC138863.1 | ||||
| BC138864.1 | ||||
| Z19542.1 | 7 | 1152 | CAA79602.1 |

Genomic Map