Assay Details
Target Gene Details
Entrez Gene ID: | 66427 |
Gene Name: | cytochrome b5 type B |
Gene Aliases: |
1810044O22Rik, AU015618, Cyb5m |
Location: |
Chr.8:107150661-107187470 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyb5b | NM_025558.5 | 5 | 3643 | NP_079834.2 |
| AK052810.1 | 5 | 3644 | BAC35156.1 | |
| AK088970.1 | 5 | 3645 | BAC40677.1 | |
| AK165543.1 | 5 | 3647 | BAE38245.1 | |
| AK168422.1 | 5 | 3633 | BAE40334.1 | |
| BC054749.1 | 5 | 3664 | AAH54749.1 | |
| BC058812.1 | 5 | 3606 | AAH58812.1 | |
| BC062980.1 | 5 | 3655 | AAH62980.1 |
More Information
Set Membership: |
|

Genomic Map