Assay Details
Target Gene Details
Entrez Gene ID: | 75767 |
Gene Name: | RAB11 family interacting protein 1 (class I) |
Gene Aliases: |
2010200K21Rik, 4833414G05Rik, Rcp |
Location: |
Chr.8:27138773-27174646 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11fip1 | NM_001080813.2 | 3 | 1016 | NP_001074282.1 |
| NM_029423.2 | 3 | 1016 | NP_083699.2 | |
| AK014696.1 | 3 | 998 | BAB29507.1 | |
| AK030769.1 | 3 | 1019 | ||
| AK086797.1 | 3 | 1014 | ||
| AK151024.1 | 3 | 1017 | BAE30042.1 | |
| BC125400.1 | 3 | 1001 | ||
| BC132094.1 | 3 | 1001 |

Genomic Map