Assay Details
Target Gene Details
Entrez Gene ID: | 75767 |
Gene Name: | RAB11 family interacting protein 1 (class I) |
Gene Aliases: |
2010200K21Rik, 4833414G05Rik, Rcp |
Location: |
Chr.8:27138773-27174646 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11fip1 | NM_001080813.2 | 6 | 4886 | NP_001074282.1 |
| NM_029423.2 | 5 | 3323 | NP_083699.2 | |
| AK131172.1 | 3 | 1817 | BAD21422.1 |

Genomic Map