Assay Details
Target Gene Details
Entrez Gene ID: | 57321 |
Gene Name: | telomeric repeat binding factor 2, interacting protein |
Gene Aliases: |
Rap1 |
Location: |
Chr.8:112011359-112020528 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Terf2ip | NM_020584.2 | 1 | 130 | NP_065609.2 |
| AB041557.1 | 1 | 84 | BAA95042.1 | |
| AB041559.1 | 1 | 85 | BAA95043.1 | |
| AK041613.1 | 1 | 76 | BAC31005.1 | |
| AK047222.1 | 1 | 87 | ||
| AK051818.1 | 1 | 50 | BAC34781.1 | |
| AK053864.1 | 1 | 86 | BAC35562.1 | |
| AK081557.1 | 1 | 86 | BAC38258.1 | |
| AK084563.1 | 1 | 78 | BAC39217.1 | |
| AK087729.1 | 1 | 88 | BAC39985.1 | |
| AK148508.1 | 1 | 90 | BAE28592.1 | |
| AK148537.1 | 1 | 92 | BAE28607.1 | |
| AK161452.1 | 1 | 108 | BAE36404.1 | |
| AK163044.1 | 1 | 86 | BAE37169.1 | |
| AK165074.1 | 1 | 88 | BAE38026.1 | |
| BC012270.1 | 1 | 46 | AAH12270.1 | |
| BC017641.1 | 1 | 106 | AAH17641.1 | |
| BY129872.1 | 1 | 130 |
More Information
Set Membership: |
|

Genomic Map