Assay Details
Target Gene Details
Entrez Gene ID: | 57321 |
Gene Name: | telomeric repeat binding factor 2, interacting protein |
Gene Aliases: |
Rap1 |
Location: |
Chr.8:112011359-112020528 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Terf2ip | NM_020584.2 | 3 | 1802 | NP_065609.2 |
| AB041557.1 | BAA95042.1 | |||
| AB041559.1 | BAA95043.1 | |||
| AK041613.1 | 3 | 1748 | BAC31005.1 | |
| AK051818.1 | 3 | 1722 | BAC34781.1 | |
| AK081557.1 | BAC38258.1 | |||
| AK084563.1 | 3 | 1750 | BAC39217.1 | |
| AK148508.1 | 3 | 1762 | BAE28592.1 | |
| AK148537.1 | 3 | 1764 | BAE28607.1 | |
| AK161452.1 | 3 | 1781 | BAE36404.1 | |
| AK165074.1 | 3 | 1760 | BAE38026.1 | |
| AK214613.1 | ||||
| AK219041.1 | ||||
| BC012270.1 | 3 | 1718 | AAH12270.1 | |
| BC017641.1 | 3 | 1778 | AAH17641.1 |
More Information
Set Membership: |
|

Genomic Map