Assay Details
Target Gene Details
Entrez Gene ID: | 546049 |
Gene Name: | RIKEN cDNA C330021F23 gene |
Gene Aliases: |
2610027C06Rik, Rps23r1, Rps23rg1 |
Location: |
Chr.8:3567998-3584776 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C330021F23Rik | NM_001024728.2 | 6 | 1467 | NP_001019899.1 |
| AK049309.1 | 6 | 1470 | BAC33674.1 | |
| AK166699.1 | 2 | 982 | ||
| BC089480.1 | 4 | 1230 |

Genomic Map