Assay Details
Target Gene Details
Entrez Gene ID: | 71544 |
Gene Name: | Rho GTPase activating protein 42 |
Gene Aliases: |
9030420J04Rik, BE136619 |
Location: |
Chr.9:8994331-9239164 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap42 | NM_027823.1 | 1 | 265 | NP_082099.1 |
| AK018520.1 | 1 | 279 | BAB31252.1 | |
| AK082857.1 | 1 | 354 | ||
| BC137891.1 | 1 | 265 | ||
| BC137892.1 | 1 | 265 |
Target Gene Details
Entrez Gene ID: | 100579151 |
Gene Name: | predicted gene, 16833 |
Gene Aliases: |
- |
Location: |
Chr.9:9236288-9260885 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16833 | NR_045754.1 | |||
| AK143934.1 | BAE25616.1 |
More Information
Set Membership: |
|

Genomic Map