Assay Details
Target Gene Details
Entrez Gene ID: | 12955 |
Gene Name: | crystallin, alpha B |
Gene Aliases: |
Crya-2, Crya2, HspB5, P23 |
Location: |
Chr.9:50751325-50756636 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cryab | NM_001289782.1 | NP_001276711.1 | ||
| NM_001289784.1 | NP_001276713.1 | |||
| NM_001289785.1 | NP_001276714.1 | |||
| NM_009964.3 | 1 | 144 | NP_034094.1 | |
| AK146556.1 | BAE27257.1 | |||
| BC010768.1 | AAH10768.1 | |||
| BC094033.1 | AAH94033.1 |

Genomic Map