Assay Details
Target Gene Details
Entrez Gene ID: | 244418 |
Gene Name: | DNA segment, Chr 8, ERATO Doi 82, expressed |
Gene Aliases: |
9830148H23, NACK, Sgk223, mFLJ00269 |
Location: |
Chr.8:36094625-36147787 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D8Ertd82e | NM_172911.3 | 3 | 1476 | NP_766499.2 |
| AK036672.1 | 3 | 1445 | BAC29528.1 | |
| AK052494.1 | 3 | 1444 | ||
| AK220205.1 | 1 | 621 | BAD90130.1 | |
| BC089024.1 | 2 | 934 | AAH89024.1 |
More Information
Set Membership: |
|

Genomic Map