Assay Details
Target Gene Details
Entrez Gene ID: | 68283 |
Gene Name: | RIKEN cDNA 9530077C05 gene |
Gene Aliases: |
1110003N12Rik, Kiaa0895, mKIAA0895 |
Location: |
Chr.9:22387732-22444684 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9530077C05Rik | NM_026739.1 | 5 | 1251 | NP_081015.1 |
| AK170604.1 | 3 | 2984 | ||
| AK210022.1 | 2 | 254 | ||
| AK220359.1 | 5 | 1315 | BAD90420.1 | |
| BC039998.1 | 3 | 474 | AAH39998.1 | |
| BC054761.1 | 5 | 1251 | AAH54761.2 |

Genomic Map