Assay Details
Target Gene Details
Entrez Gene ID: | 319278 |
Gene Name: | RIKEN cDNA A230050P20 gene |
Gene Aliases: |
- |
Location: |
Chr.9:20868592-20874307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A230050P20Rik | NM_175687.2 | 1 | 197 | NP_783618.1 |
| AK034516.1 | 1 | 104 | ||
| AK149161.1 | 1 | 177 | BAE28753.1 | |
| AK157146.1 | 1 | 182 | BAE33979.1 | |
| BC031510.1 | 1 | 209 | ||
| BC043448.1 | 1 | 143 | AAH43448.1 |

Genomic Map