Assay Details
Target Gene Details
Entrez Gene ID: | 67618 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase |
Gene Aliases: |
2010309J24Rik, 2810407B07Rik, AASD-PPT, CGI-80, LYS2, LYS5 |
Location: |
Chr.9:4294793-4309509 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aasdhppt | NM_026276.3 | 6 | 2197 | NP_080552.3 |
| AK028700.1 | 6 | 2155 | ||
| AK029523.1 | 6 | 2139 | ||
| AK031766.1 | 5 | 2019 | ||
| AK081818.1 | 6 | 2174 | ||
| AK144671.1 | 6 | 2171 | BAE26000.1 | |
| BC038013.1 | 6 | 2168 | AAH38013.1 |
More Information
Set Membership: |
|

Genomic Map