Assay Details
Target Gene Details
Entrez Gene ID: | 319583 |
Gene Name: | ligase IV, DNA, ATP-dependent |
Gene Aliases: |
5830471N16Rik, tiny |
Location: |
Chr.8:9970020-9976323 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lig4 | NM_176953.3 | 2 | 926 | NP_795927.2 |
| AK083353.1 | 2 | 803 | BAC38882.1 |
More Information
Set Membership: |
|

Genomic Map