Assay Details
Target Gene Details
Entrez Gene ID: | 76808 |
Gene Name: | ribosomal protein L18A |
Gene Aliases: |
2510019J09Rik |
Location: |
Chr.8:70894722-70897443 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl18a | NM_029751.4 | 5 | 767 | NP_084027.1 |
| AK161942.1 | 5 | 737 | BAE36645.1 |
More Information
Set Membership: |
|

Genomic Map