Assay Details
Target Gene Details
Entrez Gene ID: | 214763 |
Gene Name: | Mab-21 domain containing 1 |
Gene Aliases: |
E330016A19Rik, cGAS |
Location: |
Chr.9:78430518-78443524 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mb21d1 | NM_173386.5 | 5 | 3176 | NP_775562.2 |
| NR_125721.1 | 5 | 3220 | ||
| AK030292.1 | 5 | 3138 | BAC26881.1 | |
| AK054330.1 | 5 | 3110 | BAC35733.1 | |
| AK078662.1 | BAC37351.1 | |||
| AK145268.1 | 5 | 3116 | BAE26335.1 | |
| AK154165.1 | 5 | 3145 | ||
| BC052196.1 | 5 | 3086 |

Genomic Map