Assay Details
Target Gene Details
Entrez Gene ID: | 76491 |
Gene Name: | abhydrolase domain containing 14b |
Gene Aliases: |
1810013B01Rik |
Location: |
Chr.9:106447737-106452923 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd14b | NM_029631.3 | 4 | 1319 | NP_083907.3 |
| AK007469.1 | 4 | 1274 | ||
| BC019410.1 | 4 | 1298 | AAH19410.1 | |
| BF461776.1 | 1 | 172 |

Genomic Map