Assay Details
Target Gene Details
Entrez Gene ID: | 27358 |
Gene Name: | defensin beta 3 |
Gene Aliases: |
BD-3 |
Location: |
Chr.8:19293361-19295339 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Defb3 | NM_013756.2 | 2 | 147 | NP_038784.1 |
| AF092929.1 | 2 | 144 | AAD29572.1 | |
| BC114344.2 | 2 | 144 | AAI14345.1 | |
| BC116176.2 | 2 | 148 | ||
| BC116177.1 | 2 | 147 | ||
| BC134358.1 | 2 | 148 |
More Information
Set Membership: |
|

Genomic Map