Assay Details
Target Gene Details
Entrez Gene ID: | 71609 |
Gene Name: | TNFRSF1A-associated via death domain |
Gene Aliases: |
9130005N23Rik, AA930854 |
Location: |
Chr.8:105258575-105264594 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tradd | NM_001033161.2 | 5 | 903 | NP_001028333.1 |
| AK018592.1 | 5 | 917 | ||
| AK156540.1 | 5 | 903 | BAE33749.1 | |
| AK178733.1 | ||||
| BC132607.1 | 5 | 899 | ||
| BC138642.1 | 5 | 899 |

Genomic Map