Assay Details
Target Gene Details
Entrez Gene ID: | 11433 |
Gene Name: | acid phosphatase 5, tartrate resistant |
Gene Aliases: |
TRACP, TRAP |
Location: |
Chr.9:22126727-22135746 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acp5 | NM_001102404.1 | 4 | 757 | NP_001095874.1 |
| NM_001102405.1 | 4 | 762 | NP_001095875.1 | |
| NM_007388.3 | 4 | 704 | NP_031414.1 | |
| AK008391.1 | 3 | 484 | ||
| AK010865.1 | 4 | 728 | ||
| AK011915.1 | 4 | 728 | ||
| BC012911.1 | 4 | 656 | AAH12911.1 | |
| BC019160.1 | 4 | 720 | AAH19160.1 | |
| BC029644.1 | 4 | 656 | AAH29644.1 |
More Information
Set Membership: |
|

Genomic Map