Assay Details
Target Gene Details
Entrez Gene ID: | 11451 |
Gene Name: | acrosomal vesicle protein 1 |
Gene Aliases: |
Msa63, SP-10 |
Location: |
Chr.9:36693220-36698837 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acrv1 | NM_007391.2 | 2 | 227 | NP_031417.2 |
| AK005705.1 | 2 | 227 | BAB24197.1 | |
| AK030129.1 | 2 | 193 | ||
| BC139101.1 | 2 | 186 | ||
| BC139103.1 | 2 | 186 | ||
| U31992.1 | 2 | 198 | AAA91652.1 |

Genomic Map