Assay Details
Target Gene Details
Entrez Gene ID: | 13124 |
Gene Name: | cytochrome P450, family 8, subfamily b, polypeptide 1 |
Gene Aliases: |
- |
Location: |
Chr.9:121914356-121916305 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp8b1 | NM_010012.3 | 1 | 1623 | NP_034142.3 |
| AF090317.1 | 1 | 1609 | AAC63036.1 | |
| BC010973.1 | 1 | 1633 | AAH10973.1 | |
| BC010974.1 | 1 | 1626 | AAH10974.1 | |
| BC049969.1 | 1 | 1608 | AAH49969.1 |

Genomic Map