Assay Details
Target Gene Details
Entrez Gene ID: | 14241 |
Gene Name: | forkhead box L1 |
Gene Aliases: |
FREAC7, Fkh6, fkh-6 |
Location: |
Chr.8:121127685-121130644 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxl1 | NM_008024.2 | 1 | 2467 | NP_032050.2 |
| AK050949.1 | 1 | 1689 | BAC34473.1 |
More Information
Set Membership: |
|

Genomic Map