Assay Details
Target Gene Details
Entrez Gene ID: | 60406 |
Gene Name: | sin3 associated polypeptide |
Gene Aliases: |
30kDa |
Location: |
Chr.8:57482702-57487860 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap30 | NM_021788.2 | 1 | 508 | NP_068560.1 |
| AF075136.1 | 1 | 519 | AAC26007.1 | |
| AK010928.1 | 1 | 509 | BAB27273.1 | |
| AK088745.1 | 1 | 513 | BAC40543.1 | |
| BC132081.1 | 1 | 491 | ||
| BC132087.1 | 1 | 491 | ||
| CO800093.1 | 4 | 693 |
More Information
Set Membership: |
|

Genomic Map