Assay Details
Target Gene Details
Entrez Gene ID: | 11556 |
Gene Name: | adrenergic receptor, beta 3 |
Gene Aliases: |
Adrb-3 |
Location: |
Chr.8:27225776-27230805 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adrb3 | NM_013462.3 | 2 | 1225 | NP_038490.2 |
| AF193027.1 | 1 | 439 | AAF05768.1 | |
| AK136495.1 | 2 | 1225 | BAE23009.1 | |
| AK143775.1 | 1 | 578 | BAE25534.1 | |
| BC132000.1 | 1 | 489 |
More Information
Set Membership: |
|

Genomic Map