Assay Details
Target Gene Details
Entrez Gene ID: | 171167 |
Gene Name: | fucosyltransferase 10 |
Gene Aliases: |
fuc-TX, fut10A |
Location: |
Chr.8:31187331-31261924 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fut10 | NM_001012517.5 | 5 | 2126 | NP_001012535.1 |
| NM_001286422.1 | 6 | 2231 | NP_001273351.1 | |
| NM_001286425.1 | 4 | 1290 | NP_001273354.1 | |
| AK083041.1 | 4 | 1279 | BAC38741.1 | |
| AK147136.1 | 6 | 2187 | BAE27706.1 | |
| AK169153.1 | 5 | 2080 | BAE40932.1 |
More Information
Set Membership: |
|

Genomic Map