Assay Details
Target Gene Details
Entrez Gene ID: | 171167 |
Gene Name: | fucosyltransferase 10 |
Gene Aliases: |
fuc-TX, fut10A |
Location: |
Chr.8:31187331-31261924 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fut10 | NM_001012517.5 | 2 | 359 | NP_001012535.1 |
| NM_001286422.1 | 3 | 464 | NP_001273351.1 | |
| NM_001286424.1 | 2 | 252 | NP_001273353.1 | |
| NM_001286425.1 | 2 | 359 | NP_001273354.1 | |
| NM_134161.3 | 2 | 359 | NP_598922.1 | |
| AJ880009.1 | CAI53944.1 | |||
| AK083041.1 | 2 | 348 | BAC38741.1 | |
| AK147136.1 | 3 | 420 | BAE27706.1 | |
| AK169153.1 | 2 | 313 | BAE40932.1 | |
| BC030474.1 | 2 | 256 | AAH30474.1 | |
| BC062113.1 | 2 | 358 | AAH62113.1 | |
| BY721538.1 | 2 | 360 |
More Information
Set Membership: |
|

Genomic Map