Assay Details
Target Gene Details
Entrez Gene ID: | 171167 |
Gene Name: | fucosyltransferase 10 |
Gene Aliases: |
fuc-TX, fut10A |
Location: |
Chr.8:31187331-31261924 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fut10 | NM_001012517.5 | 4 | 1497 | NP_001012535.1 |
| NM_001286422.1 | 5 | 1602 | NP_001273351.1 | |
| NM_001286424.1 | 4 | 1390 | NP_001273353.1 | |
| NM_001286425.1 | NP_001273354.1 | |||
| NM_134161.3 | 4 | 1497 | NP_598922.1 | |
| AJ880009.1 | 3 | 1110 | CAI53944.1 | |
| AK083041.1 | BAC38741.1 | |||
| AK147136.1 | 5 | 1558 | BAE27706.1 | |
| AK169153.1 | 4 | 1451 | BAE40932.1 | |
| BC022579.1 | 1 | 595 | AAH22579.1 | |
| BC030474.1 | 4 | 1394 | AAH30474.1 | |
| BC062113.1 | 4 | 1496 | AAH62113.1 |
More Information
Set Membership: |
|

Genomic Map