Assay Details
Target Gene Details
Entrez Gene ID: | 75552 |
Gene Name: | progestin and adipoQ receptor family member IX |
Gene Aliases: |
1700020G04Rik |
Location: |
Chr.9:95559817-95562121 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paqr9 | NM_198414.2 | 1 | 1149 | NP_940806.2 |
| AK133021.1 | 1 | 1016 | BAE21473.1 | |
| AY424298.1 | 1 | 1149 | AAR08386.1 |
More Information
Set Membership: |
|

Genomic Map