Assay Details
Target Gene Details
Entrez Gene ID: | 70615 |
Gene Name: | ankyrin repeat domain 24 |
Gene Aliases: |
4631433D01Rik, 5730519E19Rik, D10Bur2e |
Location: |
Chr.10:81627795-81647612 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd24 | NM_027480.3 | 17 | 1796 | NP_081756.4 |
| AK019475.1 | 4 | 462 | BAB31744.1 | |
| AK043638.1 | 15 | 2878 | ||
| AK173316.1 | 15 | 2762 | BAD32594.1 | |
| BC017633.1 | 4 | 467 | ||
| BC048236.1 | 16 | 1701 |

Genomic Map