Assay Details
Target Gene Details
Entrez Gene ID: | 213783 |
Gene Name: | pleckstrin homology domain containing, family G (with RhoGef domain) member 1 |
Gene Aliases: |
D10Ertd733e, Gm521, mKIAA1209 |
Location: |
Chr.10:3740251-3967303 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhg1 | NM_001033253.3 | 15 | 3016 | NP_001028425.1 |
| NM_001159942.1 | 16 | 3182 | NP_001153414.1 | |
| AK122468.1 | 12 | 2340 | BAC65750.1 | |
| AK135923.1 | 15 | 3018 | BAE22726.1 | |
| BC076628.1 | 14 | 2378 | AAH76628.1 | |
| BC117965.1 | 14 | 2618 | ||
| BC117966.1 | 14 | 2611 |

Genomic Map