Assay Details
Target Gene Details
Entrez Gene ID: | 319859 |
Gene Name: | RIKEN cDNA E030011O05 gene |
Gene Aliases: |
- |
Location: |
Chr.9:96731575-96752831 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E030011O05Rik | NR_015511.2 | |||
| AK086536.1 |
Target Gene Details
Entrez Gene ID: | 245007 |
Gene Name: | zinc finger and BTB domain containing 38 |
Gene Aliases: |
A930014K01Rik, CIBZ |
Location: |
Chr.9:96685393-96731675 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb38 | NM_175537.3 | 1 | 129 | NP_780746.2 |
| AK044467.1 | 1 | 130 | BAC31937.1 |
More Information
Set Membership: |
|

Genomic Map