Assay Details
Target Gene Details
Entrez Gene ID: | 20847 |
Gene Name: | signal transducer and activator of transcription 2 |
Gene Aliases: |
1600010G07Rik, AW496480 |
Location: |
Chr.10:128270559-128292852 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat2 | NM_019963.1 | 24 | 3590 | NP_064347.1 |
| AF206162.1 | 24 | 3590 | AAF17096.1 | |
| AK005415.1 | 2 | 945 | BAB24010.2 | |
| AK030903.1 | 17 | 2682 | BAC27177.1 | |
| AK153405.1 | 22 | 3237 | BAE31965.1 | |
| AK181078.1 | ||||
| BC064827.1 | 14 | 2467 | AAH64827.1 |
More Information
Set Membership: |
|

Genomic Map