Assay Details
Target Gene Details
Entrez Gene ID: | 14426 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4 |
Gene Aliases: |
AV011803 |
Location: |
Chr.10:99108135-99113247 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt4 | NM_015737.4 | 1 | 3410 | NP_056552.1 |
| AK148036.1 | 1 | 3378 | BAE28303.1 | |
| BQ179479.1 | 1 | 656 |
Target Gene Details
Entrez Gene ID: | 382406 |
Gene Name: | POC1 centriolar protein homolog B (Chlamydomonas) |
Gene Aliases: |
4933430F16Rik, Wdr51b |
Location: |
Chr.10:99107171-99198074 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poc1b | NM_027740.6 | NP_082016.1 | ||
| AK016991.1 | BAB30542.1 | |||
| AK030180.1 | BAC26826.1 | |||
| AK045302.1 | BAC32302.1 | |||
| AK050233.1 | BAC34138.1 | |||
| AK090075.1 | BAC41080.1 | |||
| BC145801.1 |
More Information
Set Membership: |
|

Genomic Map