Assay Details
Target Gene Details
Entrez Gene ID: | 66257 |
Gene Name: | nicolin 1 |
Gene Aliases: |
1500032A17Rik |
Location: |
Chr.9:108290457-108296496 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nicn1 | NM_025449.3 | 6 | 1390 | NP_079725.1 |
| AK075732.1 | 6 | 1398 | BAC35916.1 | |
| AK146890.1 | 6 | 1400 | BAE27508.1 | |
| AK169135.1 | 6 | 1400 | BAE40914.1 | |
| BC024050.1 | 6 | 1390 | AAH24050.1 |

Genomic Map