Assay Details
Target Gene Details
Entrez Gene ID: | 13496 |
Gene Name: | AT rich interactive domain 3A (BRIGHT-like) |
Gene Aliases: |
Bright, Dri1, Dril1 |
Location: |
Chr.10:79927043-79955018 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arid3a | NM_001288625.1 | 3 | 869 | NP_001275554.1 |
| NM_001288626.1 | 3 | 1023 | NP_001275555.1 | |
| NM_007880.4 | 3 | 1023 | NP_031906.1 | |
| AK141283.1 | 3 | 870 | BAE24635.1 | |
| AK154956.1 | 3 | 995 | BAE32951.1 | |
| BC050925.1 | 3 | 927 | AAH50925.1 | |
| U60335.1 | 2 | 811 | AAB03416.1 |
More Information
Set Membership: |
|

Genomic Map