Assay Details
Target Gene Details
Entrez Gene ID: | 73690 |
Gene Name: | GLI pathogenesis-related 1 (glioma) |
Gene Aliases: |
2410114O14Rik, RTVP-1, RTVP1, mRTVP-1 |
Location: |
Chr.10:111984792-112002619 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glipr1 | NM_028608.3 | 1 | 213 | NP_082884.1 |
| AK010768.1 | 1 | 145 | BAB27168.1 | |
| AK137557.1 | 1 | 160 | BAE23406.1 | |
| AK155009.1 | 1 | 141 | ||
| AK170051.1 | 1 | 160 | BAE41532.1 | |
| BB869923.1 | 1 | 214 | ||
| BC025083.1 | 1 | 137 | AAH25083.1 | |
| BC098231.1 | 1 | 187 | AAH98231.1 |

Genomic Map