Assay Details
Target Gene Details
Entrez Gene ID: | 110012 |
Gene Name: | tubulin polyglutamylase complex subunit 1 |
Gene Aliases: |
AV005629, Gm16517, Gtrgeo22, PGs1 |
Location: |
Chr.10:79669410-79676126 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpgs1 | NM_148934.2 | 1 | 190 | NP_683736.1 |
| AF303106.1 | 1 | 190 | AAK15262.1 | |
| AK153467.1 | 1 | 117 | BAE32018.1 | |
| BC138515.1 | 1 | 231 | ||
| BC138516.1 | 1 | 231 |
More Information
Set Membership: |
|

Genomic Map