Assay Details
Target Gene Details
Entrez Gene ID: | 80907 |
Gene Name: | lactamase, beta |
Gene Aliases: |
LACT-1, Lact1, Mrpl56 |
Location: |
Chr.9:66955393-66975484 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lactb | NM_030717.1 | 1 | 370 | NP_109642.1 |
| AF317900.1 | 1 | 370 | AAG37910.1 | |
| AK078550.1 | 1 | 308 | ||
| AK087297.1 | 1 | 338 | ||
| AK149410.1 | 1 | 313 | BAE28858.1 | |
| BC016583.1 | 1 | 297 | ||
| BC021604.1 | 1 | 296 | ||
| BC046293.1 | 1 | 297 | AAH46293.1 | |
| BC147809.1 | 1 | 315 |

Genomic Map