Assay Details
Target Gene Details
Entrez Gene ID: | 80907 |
Gene Name: | lactamase, beta |
Gene Aliases: |
LACT-1, Lact1, Mrpl56 |
Location: |
Chr.9:66955393-66975484 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lactb | NM_030717.1 | 6 | 1497 | NP_109642.1 |
| AF317900.1 | 6 | 1497 | AAG37910.1 | |
| AK078550.1 | 5 | 1272 | ||
| BC046293.1 | 6 | 1424 | AAH46293.1 | |
| BC147809.1 | 6 | 1442 |

Genomic Map