Assay Details
Target Gene Details
Entrez Gene ID: | 214812 |
Gene Name: | zinc finger protein 609 |
Gene Aliases: |
9830165N24, BC016271, Znf609, mKIAA0295 |
Location: |
Chr.9:65691578-65827564 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp609 | NM_172536.3 | 6 | 3877 | NP_766124.2 |
| AK036715.1 | 5 | 3610 | BAC29549.1 | |
| AK129111.1 | 2 | 1825 | BAC97921.1 | |
| BC150683.1 | 5 | 3586 |

Genomic Map