Assay Details
Target Gene Details
Entrez Gene ID: | 22771 |
Gene Name: | zinc finger protein of the cerebellum 1 |
Gene Aliases: |
ZIC, ZNF201 |
Location: |
Chr.9:91358058-91365826 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zic1 | NM_009573.3 | 3 | 2884 | NP_033599.2 |
| AK136977.1 | 3 | 2109 | BAE23194.1 | |
| AK141431.1 | 1 | 184 | ||
| AK141883.1 | 3 | 2078 | BAE24869.1 | |
| AK159769.1 | 3 | 2856 | BAE35357.1 | |
| BC050889.1 | 3 | 2122 | ||
| BC060247.1 | 4 | 2464 | AAH60247.1 | |
| BC063247.1 | 3 | 2846 | AAH63247.1 | |
| CN666056.1 | ||||
| D32167.1 | 3 | 2632 | BAA06878.1 |
More Information
Set Membership: |
|

Genomic Map