Assay Details
Target Gene Details
Entrez Gene ID: | 76573 |
Gene Name: | spermatogenesis and centriole associated 1 like |
Gene Aliases: |
1700022B01Rik, 1700027D21Rik, AA062323 |
Location: |
Chr.10:76557552-76570201 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spatc1l | NM_029661.1 | 4 | 1065 | NP_083937.1 |
| AK006232.1 | 4 | 971 | ||
| AK006410.1 | 4 | 1065 | BAB24574.1 | |
| BC147585.1 | 4 | 1045 | ||
| BC147614.1 | 4 | 1045 |

Genomic Map