Assay Details
Target Gene Details
Entrez Gene ID: | 67537 |
Gene Name: | GLI pathogenesis-related 1 like 2 |
Gene Aliases: |
4921508O11Rik |
Location: |
Chr.10:112083344-112108098 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glipr1l2 | NM_026223.2 | 2 | 371 | NP_080499.1 |
| AK014846.1 | 2 | 372 | BAB29580.1 | |
| AK015834.1 | 2 | 355 | BAB29994.1 | |
| AK161292.1 | 2 | 372 | BAE36300.1 | |
| BC118017.1 | 2 | 372 |
More Information
Set Membership: |
|

Genomic Map