Assay Details
Target Gene Details
Entrez Gene ID: | 18646 |
Gene Name: | perforin 1 (pore forming protein) |
Gene Aliases: |
Pfn, Pfp, Prf-1 |
Location: |
Chr.10:61297680-61304263 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prf1 | NM_011073.3 | 2 | 113 | NP_035203.3 |
| BC132300.1 | 2 | 113 | ||
| BC137962.1 | 2 | 113 | ||
| M23182.1 | 2 | 252 | AAA39910.1 | |
| X12760.1 | 1 | 58 | CAA31251.1 | |
| X60165.1 | 1 | 115 | CAA42731.1 |

Genomic Map