Assay Details
Target Gene Details
Entrez Gene ID: | 108068 |
Gene Name: | glutamate receptor, metabotropic 2 |
Gene Aliases: |
4930441L02Rik, Gprc1b, mGluR2, mGluR7 |
Location: |
Chr.9:106644534-106656109 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm2 | NM_001160353.1 | 6 | 3139 | NP_001153825.1 |
| BQ174642.1 | 1 | 200 |
More Information
Set Membership: |
|

Genomic Map