Assay Details
Target Gene Details
Entrez Gene ID: | 71599 |
Gene Name: | SUMO/sentrin specific peptidase 8 |
Gene Aliases: |
9130010J17Rik, AU020827, Den1, Nedp1, Prsc2 |
Location: |
Chr.9:59734259-59750649 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Senp8 | NM_001172068.1 | 4 | 1268 | NP_001165539.1 |
| NM_001172069.1 | 3 | 1177 | NP_001165540.1 | |
| NM_001172070.1 | 3 | 1158 | NP_001165541.1 | |
| NM_001172071.1 | 3 | 1175 | NP_001165542.1 | |
| NM_027838.3 | 2 | 1065 | NP_082114.1 | |
| AK018606.1 | 3 | 1156 | BAB31304.1 | |
| AK029856.1 | 2 | 1055 | ||
| AK049120.1 | 4 | 1258 | BAC33554.1 | |
| AK136214.1 | 3 | 1148 | BAE22881.1 | |
| BC037443.1 | 2 | 1037 | AAH37443.1 |
More Information
Set Membership: |
|

Genomic Map